Learn More
Aviva Systems Biology Corp ERCC8 Antibody - C-terminal region (ARP31542_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31542_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ERCC8. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human ERCC8. Alternate names: CKN1, CSAApplicationIHC, WB. Species reactivity: Dog, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-014-54261