Learn More
Bioss ERCC8 Monoclonal Antibody
SDP
Catalog No. 5000066760
Encompass_Preferred
ERCC8 Primary Antibody Unconjugated Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB IP RecombinantConcentration 1ug/ul Size 100 ul Storage Buffer 10mM phosphate buffered saline(pH 7 4) with 150mM sodium chloride 0 05% BSA 0 02% sodium azide and 50% glycerol
Catalog No. 50-000-66760 Supplier Bioss Supplier No. BSM62393R
ERCC8 Primary Antibody Unconjugated Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB IP RecombinantConcentration 1ug/ul Size 100 ul Storage Buffer 10mM phosphate buffered saline(pH 7 4) with 150mM sodium chloride 0 05% BSA 0 02% sodium azide and 50% glycerol