Learn More
Apexbio Technology LLC Erdafitinib, 10mg CAS# 1346242-81-6
SDP
Supplier:  Apexbio Technology LLC B838710
Encompass_Preferred
Erdafitinib is an effective, selective and oral bioactive pan-fibroblast growth factor receptor FGFR inhibitor with potential anti-tumor activity and inhibits FGFR1/2/3/4 with IC50s of 1.2, 2.5, 3.0 and 5.7 nM, respectively. In vitro, Erdafitinib-treated cells reduced proliferation, increased apoptotic death, and reduced cell survival. In vivo experiments, Erdafitinib treatment delayed the growth of NCI-H716 tumors, and when drug treatment was withdrawn, tumor volume increased. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8419