Learn More
Selleck Chemical LLC Erdafitinib
SDP
Catalog No. 5000000506
Encompass_Preferred
Erdafitinib is a potent and selective orally bioavailable pan fibroblast growth factor receptor (FGFR) inhibitor with potential antineoplastic activity Erdafitinib also binds to RET (c-RET) CSF-1R PDGFR- /PDGFR- FLT4 Kit (c-Kit) and VEGFR-2 and induces cellular apoptosis
Catalog No. 50-000-00506 Supplier Selleck Chemical LLC Supplier No. S840125MG
Erdafitinib is a potent and selective orally bioavailable pan fibroblast growth factor receptor (FGFR) inhibitor with potential antineoplastic activity Erdafitinib also binds to RET (c-RET) CSF-1R PDGFR- /PDGFR- FLT4 Kit (c-Kit) and VEGFR-2 and induces cellular apoptosis