Learn More
Medchemexpress LLC HY-18708 100mg , Erdafitinib JNJ-42756493 CAS:1346242-81-6 Purity:98%
SDP
Catalog No. 501492940
Encompass_Preferred
Medchemexpress, HY-18708 100mg Erdafitinib JNJ-42756493 CAS:1346242-81-6 Formula:C25H30N6O2 IC50: 1.2 nM (FGFR1), 2.5 nM (FGFR2), 3.0 nM (FGFR3)and 5.7 nM (FGFR4) Purity:98% Erdafitinib (JNJ-42756493) is a potent and orally available FGFR family inhibitor; inhibits FGFR1-4 with IC 50 values of 1.2, 2.5, 3.0 and 5.7nM, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2940 Supplier Medchemexpress LLC Supplier No. HY18708100MG
Medchemexpress, HY-18708 100mg Erdafitinib JNJ-42756493 CAS:1346242-81-6 Formula:C25H30N6O2 IC50: 1.2 nM (FGFR1), 2.5 nM (FGFR2), 3.0 nM (FGFR3)and 5.7 nM (FGFR4) Purity:98% Erdafitinib (JNJ-42756493) is a potent and orally available FGFR family inhibitor; inhibits FGFR1-4 with IC 50 values of 1.2, 2.5, 3.0 and 5.7nM, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.