Learn More
Medchemexpress LLC HY-18708 50mg , Erdafitinib CAS:1346242-81-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1870850MG
Encompass_Preferred
HY-18708 50mg Erdafitinib CAS: 1346242-81-6 Erdafitinib (JNJ-42756493) is a potent and orally available FGFR family inhibitor; inhibits FGFR1/2/3/4 with IC50s of 1.2, 2.5, 3.0 and 5.7 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9269