Learn More
Medchemexpress LLC HY-B0289 500mg Medchemexpress, Erdosteine CAS:84611-23-4 Purity:>98%
SDP
Catalog No. 502082449
Encompass_Preferred
Medchemexpress, HY-B0289 500mg Erdosteine CAS:84611-23-4 Erdosteine inhibits lipopolysaccharide (LPS)-induced NF-κB activation. Erdosteine has muco-modulatory, anti-bacterial, anti-inflammatory and anti-oxidant effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2449 Supplier Medchemexpress LLC Supplier No. HYB0289500MG
Medchemexpress, HY-B0289 500mg Erdosteine CAS:84611-23-4 Erdosteine inhibits lipopolysaccharide (LPS)-induced NF-κB activation. Erdosteine has muco-modulatory, anti-bacterial, anti-inflammatory and anti-oxidant effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.