Learn More
ORS Nasco Erem 576TX 4 1/4 Super Full Flush Cut Magic Carbide Diagonal Tip Cutter
Catalog No. NC1431600
Encompass_Preferred
Special tapered relieved head, cutting inside .004" ID stainless wire. Super full flush cut. Handle material- cushion grip. For medical electronic applications
Catalog No. NC1431600 Supplier ORS Nasco Supplier No. 190576TX
Special tapered relieved head, cutting inside .004" ID stainless wire. Super full flush cut. Handle material- cushion grip. For medical electronic applications