Learn More
Techni Tool Inc Erem Style 15AGW Swiss Oblique Head Carbon Cutting Tweezer with Curved Tips, 4-1/2" Long
SDP
Catalog No. NC1413888
Encompass
The Erem 15AGW are 4.528" long, anti-acid, medium curved tip tweezers. The tweezers are perfect for use in micro-electronic applications.
Catalog No. NC1413888 Supplier Techni Tool Inc Supplier No. 272TW150
The Erem 15AGW are 4.528" long, anti-acid, medium curved tip tweezers. The tweezers are perfect for use in micro-electronic applications.