Learn More
AdipoGen Eremofortin A
SDP
Supplier:  AdipoGen BVT0433M001
Encompass_Preferred
Chemical. CAS 62445-06-1. Formula C17H22O5. MW 306.4. Isolated from Penicillium roqueforti. Eremophilane sesquiterpene. Nontoxic secondary metabolite from Penicillium roqueforti.
Catalog No. 50-256-1932