Learn More
Medchemexpress LLC Erenapurstat | 136164-66-4 | 378.46 | 100 MG
SDP
Catalog No. 5000385841
Encompass_Preferred
Erenapurstat is an orally active and selective inhibitor of Ape-1 (apurinic/apyrimidinic endonuclease 1)/Ref-1 (redox factor-1) redox. It has the ability to hinder tumor growth and inhibit the activity of NF-κB, AP-1, and HIF-1α in pancreatic cancer, demonstrating anticancer properties.
  • Direct, orally active and selective inhibitor of Ape-1/Ref-1 redox.
  • Impairs tumor growth.
  • Blocks the activity of NF-κB, AP-1, and HIF-1α in pancreatic cancer.
  • Shows anticancer activities.
Catalog No. 50-003-85841 Supplier Medchemexpress LLC Supplier No. HY19357100MG
Erenapurstat is an orally active and selective inhibitor of Ape-1 (apurinic/apyrimidinic endonuclease 1)/Ref-1 (redox factor-1) redox. It has the ability to hinder tumor growth and inhibit the activity of NF-κB, AP-1, and HIF-1α in pancreatic cancer, demonstrating anticancer properties.
  • Direct, orally active and selective inhibitor of Ape-1/Ref-1 redox.
  • Impairs tumor growth.
  • Blocks the activity of NF-κB, AP-1, and HIF-1α in pancreatic cancer.
  • Shows anticancer activities.