Learn More
Medchemexpress LLC Erenapurstat | 136164-66-4 | 99.67% | 378.46 | 50 MG
SDP
Catalog No. 5000385830
Encompass_Preferred
Erenapurstat (E3330) is a direct, orally active and selective inhibitor of Ape-1 (apurinic/apyrimidinic endonuclease 1)/Ref-1 (redox factor-1) redox. It impairs tumor growth and blocks the activity of NF-κB, AP-1, and HIF-1α in pancreatic cancer, showing anticancer activities.
  • Targets Ape-1 and Ref-1
  • Inhibits growth of HUVECs, PCECs, and EPCs
  • Reduces VEGF expression in pancreatic cancer cells
  • Inhibits differentiation of BM-MSCs into CD31+ endothelial lineage cells
  • Decreases cell viability in H1975 cells
  • Inhibits growth and migration of pancreatic cancer cells
  • Enhances intracellular ROS level and inhibits CD44 expression in PANC1 cells
  • Neuroprotective against cisplatin-induced alterations
  • Attenuates liver injury in rats
Catalog No. 50-003-85830 Supplier Medchemexpress LLC Supplier No. HY1935750MG
Erenapurstat (E3330) is a direct, orally active and selective inhibitor of Ape-1 (apurinic/apyrimidinic endonuclease 1)/Ref-1 (redox factor-1) redox. It impairs tumor growth and blocks the activity of NF-κB, AP-1, and HIF-1α in pancreatic cancer, showing anticancer activities.
  • Targets Ape-1 and Ref-1
  • Inhibits growth of HUVECs, PCECs, and EPCs
  • Reduces VEGF expression in pancreatic cancer cells
  • Inhibits differentiation of BM-MSCs into CD31+ endothelial lineage cells
  • Decreases cell viability in H1975 cells
  • Inhibits growth and migration of pancreatic cancer cells
  • Enhances intracellular ROS level and inhibits CD44 expression in PANC1 cells
  • Neuroprotective against cisplatin-induced alterations
  • Attenuates liver injury in rats