Learn More
Aviva Systems Biology Corp ERF Antibody - middle region (ARP32257_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32257_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ERF. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ERF. Alternate names: PE2, PE-2ApplicationWB. Species reactivity: Human
Catalog No. 50-014-54501