Learn More
Biocare Medical, LLC ERG, Concentrated and Prediluted Monoclonal Antibody, 1.0 ml
SDP
Supplier:  Biocare Medical, LLC CM421C
Encompass_Preferred
ERG has a 99.9% specificity for detecting prostatic adenocarcinomas. It does not stain lymphocytes. There is a 96.5% concordance of ERG positive PIN and ERG positive carcinoma in prostatectomy specimens. PATENT PENDING.
Catalog No. 50-827-14