Learn More
Diagnostic Biosystems ERG ERG
Catalog No. 502407475
Encompass_Preferred
ERG, the ETS related gene, belongs to the ETS family that plays important roles in cell development, differentiation, proliferation, apoptosis and tissue remodeling. The ERG antibody identifies endothelial cells, lymphocytes and prostate cancer cells.
Catalog No. 50-240-7475 Supplier Diagnostic Biosystems Supplier No. RMPD034
ERG, the ETS related gene, belongs to the ETS family that plays important roles in cell development, differentiation, proliferation, apoptosis and tissue remodeling. The ERG antibody identifies endothelial cells, lymphocytes and prostate cancer cells.