Learn More
Leica Biosystems ERG.FORCEPS 7-IN HNDL 3MM SERR
Catalog No. NC1843134
Encompass_Preferred
Available in smooth, serrated, needle tips, and with 1 x 2 teeth. The (18.5 cm) versions are for those who prefer a longer forcep and greater variety of tip choices. The curved styles are well suited for tissue separation in the water bath as well as retrieving specimens from the inside perimeter of transport jars.
Catalog No. NC1843134 Supplier Leica Biosystems Supplier No. 38DI15525
Available in smooth, serrated, needle tips, and with 1 x 2 teeth. The (18.5 cm) versions are for those who prefer a longer forcep and greater variety of tip choices. The curved styles are well suited for tissue separation in the water bath as well as retrieving specimens from the inside perimeter of transport jars.