Learn More
Medchemexpress LLC Ergi-USU-6 mesylate | 2756327-67-8 | 95.0% | C14H18N4O4S | 10MG
SDP
Catalog No. 5000260433
Encompass_Preferred
ERGi-USU-6 mesylate is a research-grade small-molecule inhibitor of ERG and RIOK2 with reported cellular potency (IC50 ≈ 0.13-0.17 μM in VCaP cells). The methanesulfonate salt is supplied as a solid with a reported purity of 95% and is intended for in vitro and preclinical research use.
  • Inhibits ERG and RIOK2 with submicromolar cellular potency.
  • Demonstrates antiproliferative activity in ERG-positive prostate cancer models.
  • Supplied as a mesylate salt for improved handling and stability.
  • High reported purity suitable for biochemical and cell-based assays.
  • Solid form with defined cold-storage recommendations for sample integrity.
Catalog No. 50-002-60433 Supplier Medchemexpress LLC Supplier No. HY13989310MG
ERGi-USU-6 mesylate is a research-grade small-molecule inhibitor of ERG and RIOK2 with reported cellular potency (IC50 ≈ 0.13-0.17 μM in VCaP cells). The methanesulfonate salt is supplied as a solid with a reported purity of 95% and is intended for in vitro and preclinical research use.
  • Inhibits ERG and RIOK2 with submicromolar cellular potency.
  • Demonstrates antiproliferative activity in ERG-positive prostate cancer models.
  • Supplied as a mesylate salt for improved handling and stability.
  • High reported purity suitable for biochemical and cell-based assays.
  • Solid form with defined cold-storage recommendations for sample integrity.