Learn More
Enzo Life Sciences ERGIC-53 monoclonal antibody (OTI1A8) (100 µl)
SDP
Supplier:  Enzo Life Sciences ENZABS3000100
Encompass_Preferred
Clone: OTI1A8. Host: Mouse. Isotype: IgG1. Immunogen: Full length recombinant human ERGIC-53. UniProt: P49257. Species Reactivity: Human, Rat, Monkey, Dog. Applications: Flow Cytometry, IF, IHC, WB. Recommended dilutions: Flow Cytometry (1:100). Immunofluorescence (1:100). Immunohistochemistry (1:150). Western Blot (1:200-1:500). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Affinity purified. Formulation: Liquid. In PBS, pH 7.3, containing 1% BSA, 50% glycerol and 0.02% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. NC1603199