Learn More
Aviva Systems Biology Corp ERGIC2 Antibody - N-terminal region (ARP39265_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39265_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ERGIon Channel2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ERGIC2. Alternate names: PTX1, CDA14, Erv41, cd002ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53132