Learn More
Aviva Systems Biology Corp ERGIC3 Antibody - C-terminal region (ARP47209_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47209_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ERGIC3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ERGIC3. Alternate names: C20orf47, CGI-54, Erv46, NY-BR-84, PRO0989, SDBCAG84, dJ477O4.2ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59486