Learn More
ACCU-SCOPE Ergo Binocular Microscope with 4X, 10X, 40XR, 50XR, 100XR oil NIS Plan achromat objective
SDP
Catalog No. 01189350 Shop All ACCU-SCOPE Products
Change view
Click to view available options
Head Style:
Binocular
Catalog No. Head Style
01-189-350 Binocular
1 options
Catalog No. 01-189-350 Supplier ACCU-SCOPE Supplier No. EXC500BEHEMA
Add to Cart
Add to Cart
Request a Quote
Delivers high-performance imaging for clinical, research, and hematology use with sharp images and adjustable controls.
  • Ergo tilting binocular viewing head 5° to 35° IPD 47 - 78 mm rotatable 360 WF 10X22 mm focusing eyepieces with eyeguards
  • Reversed sextuple nosepiece
  • 4X NIS plan 10X NIS plan 40XR NIS plan 50XR oil NIS plan 100XR oil NIS plan achromat objective
  • Hard coat mechanical stage tension adjustable controls
  • NA 0.9 Abbe condenser with iris diaphragm
  • Coaxial coarse and fine focusing controls
  • Koehler illumination 3 watt variable SLED illuminator with LIM system
  • Dust cover power cord instruction manual UL approved
Specifications
Eyepiece WF 10X/20 mm, with rubber eyeguard
Head Style Binocular
Lighting Type LED
Magnification Power 4X, 10X, 40XR, 50XR, 100XR
Model ACCU-SCOPE
Objective Types Infinity Corrected Plan
Product Type Binocular LED Microscope
Voltage 100/240 V