Learn More
ACCU-SCOPE Ergo Binocular Microscope with NIS 10X, 50X, 100X Infinity Plan Objectives
SDP
Catalog No. 01189351 Shop All ACCU-SCOPE Products
Change view
Click to view available options
Head Style:
Binocular
Catalog No. Head Style
01-189-351 Binocular
1 options
Catalog No. 01-189-351 Supplier ACCU-SCOPE Supplier No. EXC500BEMICRO
Add to Cart
Add to Cart
Request a Quote
Delivers high-performance imaging for clinical and research use with sharp, high-detail images and adjustable system controls.
  • Ergo tilting binocular viewing head 5° to 35° IPD 47 - 78 mm rotatable 360°
  • Plan achromat 10X 50XR oil and 100XR oil objectives
  • Hard coat mechanical stage tension adjustable controls
  • NA 0.9 Abbe condenser with Iris diaphragm
  • Coaxial coarse and fine focusing controls
  • Koehler illumination 3 Watt variable SLED illuminator with LIM system
  • Dust cover power cord instruction manual UL approved WF 10X22 mm focusing eyepieces with eyeguards
  • Reversed sextuple nosepiece
Specifications
Eyepiece WF 10X/20 mm, with rubber eyeguard
Head Style Binocular
Lighting Type LED
Magnification Power 10X, 50XR, 100XR
Model ACCU-SCOPE
Objective Types Infinity Corrected Plan
Product Type Binocular Compound Microscope
Voltage 100/240 V