Learn More
ACCU-SCOPE Ergo Binocular Microscope with NIS 4X, 10X, 40X Infinity Plan Objectives
SDP
Catalog No. 01189342 Shop All ACCU-SCOPE Products
Change view
Click to view available options
Head Style:
Binocular
Catalog No. Head Style
01-189-342 Binocular
1 options
Catalog No. 01-189-342 Supplier ACCU-SCOPE Supplier No. EXC500BECYT
Add to Cart
Add to Cart
Request a Quote
Supports clinical, research, and cytology workflows with high-resolution imaging and adjustable system controls.
  • Ergo tilting binocular viewing head 5° to 35° IPD 47 - 78mm rotatable 360
  • WF 10X22 mm focusing eyepieces with eyeguards
  • Reversed sextuple nosepiece
  • Plan achromat 4X 10X and 40X objectives
  • Hard coat mechanical stage tension adjustable controls
  • NA 0.9 Abbe condenser with iris diaphragm coaxial coarse and fine focusing controls Koehler illumination 3 Watt variable SLED illuminator with LIM system
  • Dust cover power cord instruction manual UL approved
Specifications
Eyepiece WF 10X/20 mm, with rubber eyeguard
Head Style Binocular
Lighting Type LED
Magnification Power 4X, 10X, 40X
Model ACCU-SCOPE
Objective Types Plan Achromatic
Product Type Binocular LED Microscope
Voltage 100/240 V