Learn More
ACCU-SCOPE™ Ergo Binocular with NIS Infinity Plan Objectives
GREENER_CHOICESDP
Catalog No. 01000982 Shop All ACCU-SCOPE Products
Change view
Click to view available options
Head Style:
Binocular, Tilting
Catalog No. Head Style
01-000-982 Binocular, Tilting
1 options
Catalog No. 01-000-982 Supplier ACCU-SCOPE™ Supplier No. EXC500BE
Add to Cart
Add to Cart
Request a Quote
EXC-500-BE delivers ergonomic excellence for prolonged use. Its ergonomic head features one-hand controls for focus, stage tension, and field diaphragm.
  • Ergo tilting binocular viewing head, 5° to 35°
  • NIS Infinity Corrected PLAN 4x, 10x, 40xR and 100xR oil objectives
  • Reversed sextuple nosepiece
  • WF 10x/22 mm focusing eyepieces with eyeguards
  • Coaxial coarse and fine focusing controls
  • Stand: Cast aluminum alloy
  • Hard coat mechanical stage, tension adjustable controls
  • N.A. 0.9 Abbe Condenser with iris diaphragm
  • Koehler illumination - 3 watt variable S-LED illuminator
  • Power Input: 100-240V 0.5A
  • Weight (approx.) 30 lb. (standard configuration with fixed binocular viewing head)
  • Dust cover, power cord, instruction manual, UL Approved
Specifications
Eyepiece WF 10X/22 mm
Head Style Binocular, Tilting
Lighting Type LED
Magnification Power 40X, 100X, 400X, 1000X
Model ACCU-SCOPE
Objective Types Infinity Corrected Plan
Product Type Binocular LED Microscope
Voltage 100/240 V