Learn More
ACCU-SCOPE™ Ergo Binocular with Plan Achromat Objectives
GREENER_CHOICESDP
Catalog No. 01000965 Shop All ACCU-SCOPE Products
Change view
Click to view available options
Head Style:
Binocular, Tilting
Catalog No. Head Style
01-000-965 Binocular, Tilting
1 options
Catalog No. 01-000-965 Supplier ACCU-SCOPE™ Supplier No. EXC360BE
Add to Cart
Add to Cart
Request a Quote
The new EXC-360-3-BE features ergonomic tilting binocular head, providing outstanding comfort during observation.
  • Ergo tilting binocular viewing head (5° to 35°)
  • Plan Achromat 4x, 10x, 40xR, 100xR oil objectives
  • PL10x/22 mm focusable eyepieces with eye cushions; accepts 24 mm reticle
  • Coaxial focusing system with adjustable tension control and upper limit stop to prevent accidental damage to slides
  • Stand: All metal structural frame with molded plastic protective shell; 5v USB-C charging port on back for personal mobile device
  • Stage: Double-layer, rackless mechanical stage with ceramic insert; size 210 x 170 mm; X-Y movement range 78 x 51 mm, Low position right hand controls
  • N.A. 1.25 Abbe Condenser with iris diaphragm; slot for phase contrast or darkfield sliders Coaxial coarse and fine focus controls
  • 3W LED with variable intensity control; LED bulb is rated for over 25,000 hours of continuous use; internal universal power supply 110-240V
  • Accessories: Dust cover, power cord, immersion oil and instruction manual
  • Warranty: 5-year limited warranty for parts, labor and electronic components; 1-year warranty on LED bulb
Specifications
Eyepiece PL-WF 10X/22 mm
Head Style Binocular, Tilting
Lighting Type LED
Magnification Power 40X, 100X, 400X, 1000X
Model ACCU-SCOPE
Objective Types Plan Achromatic
Product Type Trinocular LED Microscope
Voltage 110/240 V