Learn More
ACCU-SCOPE™ Ergo Binocular with Plan Objectives
SDP
Catalog No. 01000975 Shop All ACCU-SCOPE Products
Change view
Click to view available options
Head Style:
Binocular, Tilting
Catalog No. Head Style
01-000-975 Binocular, Tilting
1 options
Catalog No. 01-000-975 Supplier ACCU-SCOPE™ Supplier No. EXC400BE
Add to Cart
Add to Cart
Request a Quote
The EXC-400-BE is designed for a broad range of microscopy applications in clinical, academic, and research environments.
  • Optical System: Infinity optical system, f = 180 mm, anti-mold
  • Ergo tilting binocular viewing head (5° to 35°)
  • Infinity plan achromat 4x, 10x, 40xR and 100xR oil objectives
  • WF10x/22 mm focusable eyepieces with eyeguards
  • Reversed quintuple ball bearing nosepiece with positive click stops
  • Coaxial focusing, variable 3w LED illumination with field iris, dust cover
  • Stand: Cast aluminum alloy
  • Rackless mechanical stage with low-position coaxial controls; 175 x 153 mm with X-Y movement range of 76 X 50 mm
  • N.A 1.2 swing-out achromatic condenser
  • 3 Watt variable LED illuminator with variable intensity control and field iris diaphragm; LED bulb is rated for over 20,000 hours of continued use; universal power supply 110-240V
  • Warranty:5 Year limited warranty for parts, labor and electronic components 1 Year warranty on LED bulb
Specifications
Eyepiece WF 10X/22 mm
Head Style Binocular, Tilting
Lighting Type LED
Magnification Power 40X, 100X, 400X, 1000X
Model ACCU-SCOPE
Objective Types Infinity Plan Achromatic
Product Type Trinocular LED Microscope
Voltage 110/240 V