Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOTRAX™ Ergo Comfort™ Anti-Fatigue Mat

Catalog No. 19086054 Shop All NoTrax Products
Change view
Click to view available options
Length (English):
2 ft.
3 ft.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
19-086-054 3 ft.
19-086-053 2 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-086-054 Supplier NOTRAX™ Supplier No. 454S0035BL

missing translation for 'validMainframeMsg'

Ergo Comfort™ Anti-Fatigue Mat is lightweight yet comfortable

  • High quality polyurethane foam center is soft yet supportive
  • Modern diamond-top surface designed offers comfort
  • Durable, thick outer skin protects protection against wear
  • Closed cell foam construction will not absorb liquids
  • Ergonomically designed modern diamond top surface to maximize fatigue relief
  • Stain and chemical resistant surface easy to clean with common detergents and water
  • Eco-friendly polyurethane blend does not contain hazardous chemicals, plasticizers, phthalates or lead-based colors
  • Beveled edges on all four sides

Specifications

Quantity 1 Ea.
Color Black
Length (English) 3 ft.
Width (English) 5 ft.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.