Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Ergo-Fit™ Pipette
SDP

Catalog No. 12146681 Shop All Fisherbrand Products
Change view
Click to view available options
Volume (Metric):
0.2-2 μL, 2-20 μL, 20-200 μL, 100-1000 μL
1-10 μL, 10-100 μL, 100-1000 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
12-146-681 0.2-2 μL, 2-20 μL, 20-200 μL, 100-1000 μL
12-146-682 1-10 μL, 10-100 μL, 100-1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-146-681
Add to Cart
Add to Cart

Ergo-Fit contoured shape pipet with adjustable hook fits hand like a glove

  • Low force plunger enables light thumb action pipet for liquid handling
  • Plunger helps to manually control speed of aspirating and dispensing with minimal effort
  • Audible click for gentle tip acquisition and push button for tip discard
  • Cap opener at base for easy opening of PCR and microtubes
  • Simple in-lab calibration
  • Ergonomically designed pipet ensures pipetting comfort
  • Plunger allows to adjust volume for fast pipetting
  • Available in six models from 0.2 to 1000μL
  • Autoclavable pipet

Specifications

Accuracy Multiple
Autoclavable Autoclavable
Color Blue
Disposable Reusable
For Use With Pipette Tips
Grip Type Ergonomic
Material Polycarbonate ABS
Number of Channels 1
Product Line Ergo-Fit
Programmability No
Sterility Non-sterile
Type Pipette
Volume (Metric) 0.2-2 μL, 2-20 μL, 20-200 μL, 100-1000 μL
Increments Multiple
Precision Multiple
Warranty Two-year Warranty
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.