Learn More
Grainger Jamco Work Height Platform Truck, Steel, 1,200 lb. Capacity, 1 ea
Catalog No. 502790518
Encompass_Preferred
These work-height steel-deck platform trucks feature high decks designed to minimize lifting and bending when transferring items between the truck and raised surfaces such as work tables, conveyors, and storage racks. Ideal for laboratories and testing facilities, these units are suitable for transporting delicate or vibration-sensitive equipment.
  • Features shock-absorbing semi-pneumatic casters to minimize vibration
  • Equipped with a non-slip, nonconductive vinyl matted flush deck to protect items
  • Constructed from 12-gauge powder-coated steel for durability
  • Supports a load capacity of 1,200 lb
  • Includes four casters in a configuration of two rigid and two swivel wheels
Catalog No. 50-279-0518 Supplier Grainger Supplier No. 16A884
These work-height steel-deck platform trucks feature high decks designed to minimize lifting and bending when transferring items between the truck and raised surfaces such as work tables, conveyors, and storage racks. Ideal for laboratories and testing facilities, these units are suitable for transporting delicate or vibration-sensitive equipment.
  • Features shock-absorbing semi-pneumatic casters to minimize vibration
  • Equipped with a non-slip, nonconductive vinyl matted flush deck to protect items
  • Constructed from 12-gauge powder-coated steel for durability
  • Supports a load capacity of 1,200 lb
  • Includes four casters in a configuration of two rigid and two swivel wheels