Learn More
ACCU-SCOPE™ Ergo Trinocular with NIS Infinity Plan Objectives
SDP
Catalog No. 01000984 Shop All ACCU-SCOPE Products
Change view
Click to view available options
Head Style:
Trinocular, Tilting
Catalog No. Head Style
01-000-984 Trinocular, Tilting
1 options
Catalog No. 01-000-984 Supplier ACCU-SCOPE™ Supplier No. EXC500TE
Add to Cart
Add to Cart
Request a Quote
EXC-500-TE features an ergonomic trinocular head (0° to 35° adjustable, IPD 47-78 mm, 360° rotation), paired with NIS Infinity Plan 4x, 10x, 40xR,100xR objectives.
  • Ergo Trinocular viewing head, adjustable 0° to 35°, IPD 47 - 78 mm, rotatable 360°
  • Viewing split: 100% T / 0% B; 20% T / 80% B; and 0% T / 100% B
  • NIS Infinity Corrected PLAN 4x, 10x, 40xR and 100xR oil objectives
  • Reversed sextuple nosepiece
  • WF 10x/22 mm focusing eyepieces with eyeguards
  • Coaxial coarse and fine focusing controls
  • Stand: Cast aluminum alloy
  • Hard coat mechanical stage, tension adjustable controls
  • N.A. 0.9 Abbe Condenser with iris diaphragm
  • Koehler illumination - 3 watt variable S-LED illuminator
  • Power Input: 100-240V 0.5A
  • Dust cover, power cord, instruction manual, UL Approved
Specifications
Eyepiece WF 10X/22 mm
Head Style Trinocular, Tilting
Lighting Type LED
Magnification Power 40X, 100X, 400X, 1000X
Model ACCU-SCOPE
Objective Types Infinity Corrected Plan
Product Type Trinocular LED Microscope
Voltage 100/240 V