Learn More
METTLER TOLEDO™ ErgoClip Tare Container Holder for Micro-Analytical Balances
Catalog No. 13940067 Shop All METTLER TOLEDO Products
Change view
Click to view available options
ErgoClip Tare Container Holder for Micro-Analytical Balances
Catalog No.
13-940-067 ErgoClip Tare Container Holder for Micro-Analytical Balances
1 options
Catalog No. 13-940-067 Supplier METTLER TOLEDO™ Supplier No. 30078848
Add to Cart
Add to Cart
Direct Substance Dosing
  • Weigh Directly into Tare Containers
  • ErgoClips micro can be easily assembled on XP26 and XP56 Micro Balances allowing one step dosing directly into the tare container
  • Fast and Safe Dosing
  • rgoClips safer dosing method substantially reduces contamination risks, enables fast dosing processes and increases productivity
  • Accurate Weighing
  • Dosing directly into the tare container eliminates transfer errors and saves money by minimizing sample loss.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) XPE26/56 Micro Balance
Quantity 1
Type Tare Container Holder for PCR, Eppendorf or Reaction Tubes