Learn More
METTLER TOLEDO™ ErgoClip Tare Container Holder for Micro Balances
Catalog No. 13940066 Shop All METTLER TOLEDO Products
Change view
Click to view available options
ErgoClip Tare Container Holder
Catalog No.
13-940-066 ErgoClip Tare Container Holder
1 options
Catalog No. 13-940-066 Supplier METTLER TOLEDO™ Supplier No. 11107879
Add to Cart
Add to Cart
Direct Substance Dosing
Tare container holder for 1 to 10 mL volumetric flasks to use on XPE26/56 micro balances
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) XPE26/56 Micro Balance
For Use With (Application) Direct substance dosing
Quantity 1
Type Tare Container Holder for 1 to 10 mL Volumetric Flasks