Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wearwell™ ErgoDeck Diamond Plate General Purpose Solid Matting

Catalog No. 19060521 Shop All Wearwell Tennessee Mat Company Inc. Products
Change view
Click to view available options
Color:
Black
Charcoal
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-060-521 Charcoal
19-060-520 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-060-521 Supplier Wearwell™ Supplier No. 590.78X18X18CHCS10
  • $936.00
    $559.65 / Case of 10

    Save $376.35

    40% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Wearwell™ ErgoDeck with Diamond-Plate has a box-spring foundation that compresses under foot, providing anti-fatigue benefits plus the necessary structure to support heavy foot traffic

  • Standing relief in wet or dry applications
  • Customize in 3-inch increments to fit any area or configuration
  • Positive Interlocking System prevents separation
  • Easily replace single pieces or sections as needed to reduce costs
  • 100% Silicone Free PVC tiles provide maximum durability
  • Diamond-Plate surface pattern delivers dependable traction and offers an Industrial look

Specifications

Color Charcoal
Thickness (English) 7/8 in.
Dimensions (L x W) 18 x 18 in.
Thickness (Metric) 22 mm
For Use With (Application) Oily areas
Surface Diamond plate pattern
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.