Description
- Highly accurate powder filling into capsules
- Ideal for small batches up to 12 capsules at a time
- Increased productivity
Specifications
Specifications
| For Use With (Equipment) | Excellence XPR Automatic Balance |
| Product Line | ErgoDisc Capsule Holder |
| Type | Balance Capsule Holder |