Learn More
METTLER TOLEDO™ ErgoDisc Capsule Holder
Catalog No. 01677040 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Size:
0
00
1
2
3
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
01-677-040 0
01-677-011 -
01-677-036 2
01-677-037 1
01-677-038 00
01-677-039 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 01-677-040 Supplier METTLER TOLEDO™ Supplier No. 11141751
Only null left
Add to Cart
Add to Cart
Dosing accessories enable faster dosing of more capsules in less time to increase throughput and productivity in your daily processes.
  • Highly accurate powder filling into capsules
  • Ideal for small batches up to 12 capsules at a time
  • Increased productivity
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Excellence XPR Automatic Balance
Size 0
Product Line ErgoDisc Capsule Holder
Type Balance Capsule Holder