Learn More
MSC Ergodyne Anti-Vibration Gloves, ProFlex 9012, Medium, Wrist Support, 1 Pair
Catalog No. 5000801959
Encompass
These certified anti-vibration gloves offer robust hand protection and wrist support for general-purpose applications. They effectively reduce vibration and impact while providing comfort for repetitive tasks, ensuring a secure grip and compliance with key industry standards.
  • ANSI/ISEA 105-2016, ANSI/ASA S2.73-2014/ISO 10819:2013, EN 388 certified anti-vibration protection
  • Full-length AVC palm padding reduces vibration and palm impact
  • Built-in wrist support with patented Open-Center Stay aids in relief from repetitive motion discomfort
  • Gripping padded palm provides secure hold on tools and equipment
  • Technical hex stretch knit construction and neoprene knuckle pad
  • Reinforced fingertips and thumb saddle
  • Low-profile molded closure tab with ID space
Catalog No. 50-008-01959 Supplier MSC Supplier No. 38334041
These certified anti-vibration gloves offer robust hand protection and wrist support for general-purpose applications. They effectively reduce vibration and impact while providing comfort for repetitive tasks, ensuring a secure grip and compliance with key industry standards.
  • ANSI/ISEA 105-2016, ANSI/ASA S2.73-2014/ISO 10819:2013, EN 388 certified anti-vibration protection
  • Full-length AVC palm padding reduces vibration and palm impact
  • Built-in wrist support with patented Open-Center Stay aids in relief from repetitive motion discomfort
  • Gripping padded palm provides secure hold on tools and equipment
  • Technical hex stretch knit construction and neoprene knuckle pad
  • Reinforced fingertips and thumb saddle
  • Low-profile molded closure tab with ID space