Learn More
MSC Ergodyne Anti-Vibration Gloves, ProFlex 9012, Wrist Support, X-Large, 1 ea
Catalog No. 5000794298
Encompass
These Certified Anti-Vibration Gloves + Wrist Support feature full-length AVC palm padding to reduce vibration. A built-in wrist support with an Open-Center Stay aids in relief from repetitive motion discomfort. The gloves meet ANSI/ISEA 105-2016, ANSI/ASA S2.73-2014/ISO 10819:2013, and EN 388 anti-vibration protection standards.
  • ANSI/ISEA 105-2016, ANSI/ASA S2.73-2014/ISO 10819:2013, EN 388 anti-vibration standards
  • Full-length AVC palm padding reduces vibration and palm impact
  • Built-in wrist support with patented open-center stay for repetitive motion discomfort
  • Gripping palm provides secure hold on tools and equipment
  • Hex stretch knit construction
  • Neoprene knuckle pad
  • Reinforced fingertips and thumb saddle
  • Low-profile molded closure tab with ID space to write wearer's name
  • EN 388: 2121X
  • Patent pending
Catalog No. 50-007-94298 Supplier MSC Supplier No. 38714978
These Certified Anti-Vibration Gloves + Wrist Support feature full-length AVC palm padding to reduce vibration. A built-in wrist support with an Open-Center Stay aids in relief from repetitive motion discomfort. The gloves meet ANSI/ISEA 105-2016, ANSI/ASA S2.73-2014/ISO 10819:2013, and EN 388 anti-vibration protection standards.
  • ANSI/ISEA 105-2016, ANSI/ASA S2.73-2014/ISO 10819:2013, EN 388 anti-vibration standards
  • Full-length AVC palm padding reduces vibration and palm impact
  • Built-in wrist support with patented open-center stay for repetitive motion discomfort
  • Gripping palm provides secure hold on tools and equipment
  • Hex stretch knit construction
  • Neoprene knuckle pad
  • Reinforced fingertips and thumb saddle
  • Low-profile molded closure tab with ID space to write wearer's name
  • EN 388: 2121X
  • Patent pending