Learn More
MSC Ergodyne Arm Sleeves, A5 Cut Resistant, 22 in, 1 Pair
Catalog No. 5000792051
Encompass
ProFlex 7951 Cut-Resistant Protective Arm Sleeves use aramid fibers for exceptional cut (ANSI/ISEA 105-2016 A5) and heat (EN 407 level 1, up to 212°F/100°C for 15s) protection. Designed for comfort, they ensure natural movement and prevent exposure to hazards.
  • Includes two sleeves for left and right arms.
  • ANSI/ISEA 105-2016 A5 cut protection for sharp material handling.
  • Aramid fiber provides EN 407 level 1 heat protection.
  • Adjustable hook & loop strap ensures snug upper arm/bicep fit.
  • Soft neoprene thumb loops keep sleeve in place.
  • Pre-curved design and flexible elbow zones for mobility.
  • Machine washable; meets ANSI/ISEA 105-2016 A5 cut, level 3 abrasion, EN 388: 2X4X F, EN407 level 1 standards.
Catalog No. 50-007-92051 Supplier MSC Supplier No. 79036620
ProFlex 7951 Cut-Resistant Protective Arm Sleeves use aramid fibers for exceptional cut (ANSI/ISEA 105-2016 A5) and heat (EN 407 level 1, up to 212°F/100°C for 15s) protection. Designed for comfort, they ensure natural movement and prevent exposure to hazards.
  • Includes two sleeves for left and right arms.
  • ANSI/ISEA 105-2016 A5 cut protection for sharp material handling.
  • Aramid fiber provides EN 407 level 1 heat protection.
  • Adjustable hook & loop strap ensures snug upper arm/bicep fit.
  • Soft neoprene thumb loops keep sleeve in place.
  • Pre-curved design and flexible elbow zones for mobility.
  • Machine washable; meets ANSI/ISEA 105-2016 A5 cut, level 3 abrasion, EN 388: 2X4X F, EN407 level 1 standards.