Learn More
MSC Ergodyne Bump Cap, Adjustable, Vented, Nylon, 1 ea
Catalog No. 5000687004
Encompass
This bump cap provides head protection in environments where there is a risk of bumping the head on stationary objects. It features a lightweight design for comfort and compliance with EN 812 standards.
  • Adjustable hook and loop strap
  • Brim size of 80 mm
  • Vented for breathability
  • Made from durable nylon material
  • Available in navy color
  • Compliant with EN 812 safety standards
Catalog No. 50-006-87004 Supplier MSC Supplier No. 40532558
This bump cap provides head protection in environments where there is a risk of bumping the head on stationary objects. It features a lightweight design for comfort and compliance with EN 812 standards.
  • Adjustable hook and loop strap
  • Brim size of 80 mm
  • Vented for breathability
  • Made from durable nylon material
  • Available in navy color
  • Compliant with EN 812 safety standards