Learn More
MSC Ergodyne Bump Cap Hat, LED Lighting, Black, 1 ea
Catalog No. 5000803187
Encompass
The Skullerz 8960 Bump Cap Hat provides comfortable protection against minor head injuries. It features integrated LED lights for hands-free visibility and a removable impact-resistant ABS plastic shell with foam cushioning. The design promotes breathability and includes reflective accents for enhanced worker safety.
  • Guards against minor head injuries
  • Built-in LED lights provide 48 lumens for 30 hours of hands-free visibility
  • Removable impact-resistant ABS plastic shell with foam lining
  • Flanged design maximizes airflow for breathability
  • Reflective piping enhances worker visibility
  • Short 50 mm brim for upward visibility and shade
  • Meets ANSI/ISEA 100-2024 & EN 812-2012 standards for industrial bump caps
  • Washable design with removable batteries and insert
  • One size fits most (accommodates head sizes up to 7 ½)
Catalog No. 50-008-03187 Supplier MSC Supplier No. 40532491
The Skullerz 8960 Bump Cap Hat provides comfortable protection against minor head injuries. It features integrated LED lights for hands-free visibility and a removable impact-resistant ABS plastic shell with foam cushioning. The design promotes breathability and includes reflective accents for enhanced worker safety.
  • Guards against minor head injuries
  • Built-in LED lights provide 48 lumens for 30 hours of hands-free visibility
  • Removable impact-resistant ABS plastic shell with foam lining
  • Flanged design maximizes airflow for breathability
  • Reflective piping enhances worker visibility
  • Short 50 mm brim for upward visibility and shade
  • Meets ANSI/ISEA 100-2024 & EN 812-2012 standards for industrial bump caps
  • Washable design with removable batteries and insert
  • One size fits most (accommodates head sizes up to 7 ½)