Learn More
Ergodyne™ Chill-Its™ 6705 Bandana/Headband
Catalog No. 19058454 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Color:
Navy Western
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-058-454 Navy Western
19-120-2705 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-058-454 Supplier Ergodyne™ Supplier No. 12317
This item is not returnable. View return policy
Evaporative cooling process keeps you cool
Provides cool comfort for up to 4 hours when activated in cold water
Ideal for all warm work—helps workers stay cool
  • Hook and loop closure for easy on and off
  • One size fits all
  • Reusable: just resoak
  • Guaranteed against manufactured defects
  • Hand wash
Specifications
Product Type Bandana/Headband
Size Universal
Color Navy Western
For Use With (Application) Construction, trades, maintenance, landscaping/grounds, assembly/fabrication, material handling, freight/baggage, warehousing/distribution, Iron/Steel fabrication
Includes Hook and Loop Closure