Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Chill-Its™ 6700 Evaporative Cooling Tie Bandana

Catalog No. 19058452 Shop All Ergodyne Products
Change view
Click to view available options
Color:
Navy
Red
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-058-452 Navy
19-058-459 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-058-452 Supplier Ergodyne™ Supplier No. 12306
Add to Cart
Add to Cart

Cool comfort for 24 to 48 hours when activated in cool water

Ideal for workers in construction, landscaping, foundries, mining, welding, factories, farming and food service.

  • Evaporative cooling works all day
  • Lightweight, comfrotable cotton construction provides all-day comfort
  • Super absorbent acrylic polymer activates easily when soaked in water for 15 mins.
  • Machine-washable; Reusable

Specifications

Size Universal
Color Navy
For Use With (Application) Ideal for workers in construction, landscaping, foundries, mining, welding, factories, farming and food service
Includes Activated Polymers, Tie Closure
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.