Learn More
Description
- Hi Cool™ Moisture Management performance fabric
- Comfortable to wear under hats, helmets, or alone
- Wide, absorbent terrycloth band
- Elastic band for a great fit
Specifications
Specifications
| Product Type | High Performance Cap |
| Color | Stars and Stripes |
| Band Type | Wide, Absorbent Terrycloth |
| For Use With (Application) | Construction, trades, maintenance, landscaping/grounds, assembly/fabrication, material handling, freight/baggage, warehousing/distribution, Iron/Steel fabrication, hells kitchen |
| For Use With (Equipment) | For use under hats, helmets, or alone |
| Includes | Elastic Band |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.