Learn More
MSC Ergodyne Cooling Vest, 38 to 42 in. Chest, 1 ea
Catalog No. 5000674287
Encompass
This evaporative cooling vest is designed to provide cooling relief, featuring a durable nylon construction and a convenient zipper front. It helps reduce heat stress in various environments by activating simply with cold water.
  • Evaporative cooling technology provides heat stress relief.
  • Lightweight, durable, and washable construction.
  • Contains cooling crystals for effective cooling.
  • Water-repellent liner helps keep you dry.
  • Activates by soaking in cold water for 2-5 minutes.
  • Provides up to 4 hours of maximum cooling.
  • Made from nylon material.
  • Features a convenient zipper closure.
  • Size large with a lime color.
Catalog No. 50-006-74287 Supplier MSC Supplier No. 79115101
This evaporative cooling vest is designed to provide cooling relief, featuring a durable nylon construction and a convenient zipper front. It helps reduce heat stress in various environments by activating simply with cold water.
  • Evaporative cooling technology provides heat stress relief.
  • Lightweight, durable, and washable construction.
  • Contains cooling crystals for effective cooling.
  • Water-repellent liner helps keep you dry.
  • Activates by soaking in cold water for 2-5 minutes.
  • Provides up to 4 hours of maximum cooling.
  • Made from nylon material.
  • Features a convenient zipper closure.
  • Size large with a lime color.