Learn More
MSC Ergodyne Cooling Vest, Phase Change, S/M, Khaki, 1 ea
Catalog No. 5000788760
Encompass
The Chill-Its 6230 Standard Phase Change Cooling Vest, with rechargeable packs, provides immediate and long-lasting cooling. It maintains a constant 64°F (18°C) for up to 4 hours, making it ideal for hot indoor and outdoor work environments or for wearing under PPE.
  • Immediate and long-lasting cooling relief, maintaining 64°F (18°C) for up to 4 hours.
  • Easy activation: packs recharge quickly in ice water, refrigerator, freezer, or near AC.
  • Honeycomb design eliminates stiffness for comfort and flexibility.
  • Adjustable hook and loop closures on shoulders and waist ensure a customized fit.
  • Extended vest surface area enhances cooling power.
  • Made from comfortable cotton blend fabric; vest is machine washable.
Catalog No. 50-007-88760 Supplier MSC Supplier No. 31826936
The Chill-Its 6230 Standard Phase Change Cooling Vest, with rechargeable packs, provides immediate and long-lasting cooling. It maintains a constant 64°F (18°C) for up to 4 hours, making it ideal for hot indoor and outdoor work environments or for wearing under PPE.
  • Immediate and long-lasting cooling relief, maintaining 64°F (18°C) for up to 4 hours.
  • Easy activation: packs recharge quickly in ice water, refrigerator, freezer, or near AC.
  • Honeycomb design eliminates stiffness for comfort and flexibility.
  • Adjustable hook and loop closures on shoulders and waist ensure a customized fit.
  • Extended vest surface area enhances cooling power.
  • Made from comfortable cotton blend fabric; vest is machine washable.