Learn More
MSC Ergodyne Glove Belt Clip, Breakaway Design, Acetal Copolymer, 1 ea
Catalog No. 5000767761
Encompass
This glove clip holder with a belt clip is designed to keep gloves, PPE, and other items conveniently at hand. It attaches to a belt or harness and features a patented breakaway design for safety, allowing reassembly if detached. Constructed from durable, non-conductive, and chemical-resistant acetal copolymer, it securely holds bulky items.
  • Keeps gloves, PPE, and towels within reach.
  • Features a strong gripping clip that is easy to open and stays firmly closed.
  • Constructed from durable, ultra-resilient acetal copolymer and thermoplastic polyurethane.
  • Includes a patented breakaway design that detaches if caught, allowing reassembly.
  • Helps prevent dropped and lost objects.
  • Average breaking strength of 26 lb (11.8 kg).
  • Measures 6 in (15.2 cm) long x 1 in (2.5 cm) wide x 1 in (2.5 cm).
  • Made in the USA.
Catalog No. 50-007-67761 Supplier MSC Supplier No. 45647286
This glove clip holder with a belt clip is designed to keep gloves, PPE, and other items conveniently at hand. It attaches to a belt or harness and features a patented breakaway design for safety, allowing reassembly if detached. Constructed from durable, non-conductive, and chemical-resistant acetal copolymer, it securely holds bulky items.
  • Keeps gloves, PPE, and towels within reach.
  • Features a strong gripping clip that is easy to open and stays firmly closed.
  • Constructed from durable, ultra-resilient acetal copolymer and thermoplastic polyurethane.
  • Includes a patented breakaway design that detaches if caught, allowing reassembly.
  • Helps prevent dropped and lost objects.
  • Average breaking strength of 26 lb (11.8 kg).
  • Measures 6 in (15.2 cm) long x 1 in (2.5 cm) wide x 1 in (2.5 cm).
  • Made in the USA.