Learn More
Enterprise Technology Solutions Ergodyne Gloves,Nitrile-Coated,ProFlex,Small,7"-8",12PR/BG,BK
SDP
Catalog No. 5000828409
Encompass_Preferred
ProFlex 7001 Nitrile-Coated Gloves with DSX Dip technology provide excellent grip in dry conditions. Close, second-skin fit is made for top-of-the-line dexterity and comfort when working with small parts in manufacturing or assembly. This soft, flexible fit is thanks to 18-gauge, seamless, ANSI level 3, abrasion-resistant nylon plus spandex knit blend. Thin knit allows unmatched breathability and a fit like no other. DSX Nitrile Dip technology outperforms all nitrile-based dips in dry environments. Multi-layer dip allows air to flow through to keep hands comfortable and dry. Top-layer dip creates a highly abrasion-resistant coating for durability and longer life. Touch screen-capable fingertips allow workers to stay connected and protected while maximizing productivity. These machine-washable safety gloves are best for workers in assembly, manufacturing, automotive and service technicians and are perfect for applications involving small parts or tasks requiring dexterity.
Catalog No. 50-008-28409 Supplier Enterprise Technology Solutions Supplier No. EGO17952
ProFlex 7001 Nitrile-Coated Gloves with DSX Dip technology provide excellent grip in dry conditions. Close, second-skin fit is made for top-of-the-line dexterity and comfort when working with small parts in manufacturing or assembly. This soft, flexible fit is thanks to 18-gauge, seamless, ANSI level 3, abrasion-resistant nylon plus spandex knit blend. Thin knit allows unmatched breathability and a fit like no other. DSX Nitrile Dip technology outperforms all nitrile-based dips in dry environments. Multi-layer dip allows air to flow through to keep hands comfortable and dry. Top-layer dip creates a highly abrasion-resistant coating for durability and longer life. Touch screen-capable fingertips allow workers to stay connected and protected while maximizing productivity. These machine-washable safety gloves are best for workers in assembly, manufacturing, automotive and service technicians and are perfect for applications involving small parts or tasks requiring dexterity.