Learn More
MSC Ergodyne Gloves, ProFlex 9000 Series, Anti-Vibration, Large, Cotton/Nylon, 1 Pair
Catalog No. 5000792032
Encompass
These anti-vibration gloves protect hands from continuous vibration exposure from power tools and machinery. They feature a unique chloroprene rubber palm pad and a lightweight 7-gauge cotton/nylon seamless knit for comfort, dexterity, and breathability. A pre-curved design reduces hand fatigue, and an elastic cuff prevents debris entry.
  • Anti-vibration protection from chloroprene rubber palm and fingers
  • Ergonomic pre-curved design minimizes hand fatigue
  • Flexible fit with breathable 7-gauge cotton/nylon seamless knit back
  • Elastic cuff for secure fit and debris prevention
  • Meets EN388:2016 and ISO 10819:2013 anti-vibration standards
Catalog No. 50-007-92032 Supplier MSC Supplier No. 73376352
These anti-vibration gloves protect hands from continuous vibration exposure from power tools and machinery. They feature a unique chloroprene rubber palm pad and a lightweight 7-gauge cotton/nylon seamless knit for comfort, dexterity, and breathability. A pre-curved design reduces hand fatigue, and an elastic cuff prevents debris entry.
  • Anti-vibration protection from chloroprene rubber palm and fingers
  • Ergonomic pre-curved design minimizes hand fatigue
  • Flexible fit with breathable 7-gauge cotton/nylon seamless knit back
  • Elastic cuff for secure fit and debris prevention
  • Meets EN388:2016 and ISO 10819:2013 anti-vibration standards