Learn More
MSC Ergodyne GloWear 8915 Pants, Class E, Orange, Small, 1 ea
Catalog No. 5000765813
Encompass
The GloWear 8915 Hi-Vis Rain Pants offer waterproof, breathable protection against wet conditions. Constructed with durable 300D poly oxford material and a breathable polyurethane (PU) coating, these pants ensure comfort by allowing moisture to escape while keeping rain out. They meet ANSI/ISEA 107-2020 Supplemental Class E Standards and are designed for industrial workplaces requiring enhanced visibility.
  • Waterproof and rugged 300D oxford polyester outer shell
  • Durable and breathable PU coating
  • Two in. reflective material compliant to ANSI/ISEA 107-2020 for nighttime visibility
  • Comfortable elastic waistband with drawstring
  • Zippered leg openings for easy on/off
  • Two front side pockets and one right leg cargo pocket
Catalog No. 50-007-65813 Supplier MSC Supplier No. 70448741
The GloWear 8915 Hi-Vis Rain Pants offer waterproof, breathable protection against wet conditions. Constructed with durable 300D poly oxford material and a breathable polyurethane (PU) coating, these pants ensure comfort by allowing moisture to escape while keeping rain out. They meet ANSI/ISEA 107-2020 Supplemental Class E Standards and are designed for industrial workplaces requiring enhanced visibility.
  • Waterproof and rugged 300D oxford polyester outer shell
  • Durable and breathable PU coating
  • Two in. reflective material compliant to ANSI/ISEA 107-2020 for nighttime visibility
  • Comfortable elastic waistband with drawstring
  • Zippered leg openings for easy on/off
  • Two front side pockets and one right leg cargo pocket