Learn More
MSC Ergodyne Hi-Vis Safety Vest, Class 2, 1 ea
Catalog No. 5000686337
Encompass
The GloWear 8205HL Hi-Vis Safety Vest provides enhanced visibility in low-light and nighttime conditions. It is ANSI/ISEA 107-2020, Type R, Class 2 compliant, featuring hi-vis fabric and 2-inch reflective tape for safety. Constructed from lightweight 3.3oz polyester mesh, the vest ensures comfort throughout the day.
  • Increased visibility: ANSI/ISEA 107-2020, Type R, Class 2 compliant with 2 in reflective tape
  • All-day comfort: Made from 3.3 oz polyester mesh fabric
  • Easy on and off: Features a hook and loop closure for adjustability
  • Easy maintenance: Machine-washable fabric retains shape for up to 25 washes
Catalog No. 50-006-86337 Supplier MSC Supplier No. 45750650
The GloWear 8205HL Hi-Vis Safety Vest provides enhanced visibility in low-light and nighttime conditions. It is ANSI/ISEA 107-2020, Type R, Class 2 compliant, featuring hi-vis fabric and 2-inch reflective tape for safety. Constructed from lightweight 3.3oz polyester mesh, the vest ensures comfort throughout the day.
  • Increased visibility: ANSI/ISEA 107-2020, Type R, Class 2 compliant with 2 in reflective tape
  • All-day comfort: Made from 3.3 oz polyester mesh fabric
  • Easy on and off: Features a hook and loop closure for adjustability
  • Easy maintenance: Machine-washable fabric retains shape for up to 25 washes